Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ AZD 2858

Click to view available options
Quantity:
10 mg
50 mg
Description
AZD 2858 is a potent glycogen synthase kinase-3 (GSK-3) inhibitor (IC50 values are 0.9 and 5 nM for GSK-3α and GSK-3β, respectively). AZD 2858 inhibits tau phosphorylation in vitro (IC50 = 76 nM), improves fracture healing and increases bone mass in rodent models of bone fracture. Also activates Wnt signaling in vitro and in vivo. Brain permeable and orally bioavailable.
Specifications
Specifications
Chemical Name or Material | 3-Amino-6-[4-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-N-3-pyridinyl-2-pyrazinecarboxamide |
CAS | 486424-20-8 |
Quantity | 10 mg |
Target | Glycogen Synthase Kinase Inhibitors |
Molecular Formula | C21H23N7O3S |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction