Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 5363
GREENER_CHOICE

Catalog No. p-7166835
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent pan-AKT inhibitor

AZD 5363 is a potent pan-AKT inhibitor (IC50 values are 3, 7 and 7 nM for AKt 1, 2 and 3, respectively). Also inhibits P70S6K, PKA, ROCK2 and ROCK1 (IC50 values are 6, 7, 60 and 470 nM, respectively). Inhibits growth of tumor cell lines in vitro, particularly breast, endometrial, gastric, hematologic and prostate cancers. Inhibits growth of human tumor xenografts in mice. Enhances in vivo antitumor effects of Docetaxel (Cat. No. 4056) and Lapatinib (Cat. No. 6811). Orally bioavailable.

Specifications

Synonym Capivasertib
Quantity 5 mg
Molecular Formula C21H25ClN6O2
Formula Weight 428.92
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.