Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 5582 dihydrochloride
GREENER_CHOICE

Catalog No. p-5545701
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Dimeric Smac mimetic; potent IAP inhibitor

AZD 5582 dihydrochloride is a dimeric Smac mimetic; potent inhibitor of X-linked (XIAP) and cellular (cIAP) inhibitor of apoptosis protein (IC50 values are 15, 15 and 21 nM for XIAP, cIAP1 and cIAP2 respectively). Binds to the BIR3 domain of XIAP to prevent interaction with caspase-9. Causes degradation of cIAP1 and cIAP2 and induces apoptosis in MDA-MB-231 breast cancer cells. Causes tumor regression in MDA-MB-231 xenograft-bearing mice.

Specifications

Quantity 10 mg
Product Type AZD 5582 dihydrochloride
Purity >97%
Molecular Weight (g/mol) 1088.21
Inhibitors AZD 5582 dihydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.