Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 6482
GREENER_CHOICESDP

Catalog No. 483910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-391-0 10 mg
48-395-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-391-0 Supplier Tocris Bioscience™ Supplier No. 4839/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PI 3-Kβ inhibitor

Potent and selective inhibitor of PI 3-kinase β (PI 3-Kβ) (IC50 values are 0.69, 13.6, 47.8 and 136 nM for PI 3-Kβ, PI 3-Kδ, PI 3-Kγ and PI 3-Kα, respectively). Selectively inhibits in vivo growth of the PTEN-deficient tumor xenografts HCC70 and PC3. Also exhibits antithrombotic activity.

Chemical Identifiers

CAS 1173900-33-8
Molecular Formula C22H24N4O4

Specifications

Quantity 10 mg
Formula Weight 408.45
Purity >98%
Molecular Weight (g/mol) 408.458
Chemical Name or Material AZD 6482
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.