Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Azithromycin
GSA_VAGREENER_CHOICESDP

Catalog No. 377150 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 37-715-0 Supplier R&D Systems™ Supplier No. 3771/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antibiotic; inhibits 50S ribosomal subunit formation and elongation at transpeptidation

Azithromycin is a macrolide antibiotic. Inhibits 50S ribosomal subunit formation and elongation at transpeptidation step in gram-positive and gram-negative organisms. Orally active with improved pharmacokinetics over erythromycin in mouse models. Inhibits autophagy. Predicted to disrupt binding of SARS-CoV-2 spike protein to ACE2.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 83905-01-5
Molecular Formula C38H72N2O12

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 748.98
Product Type Azithromycin
Purity 99%
Molecular Weight (g/mol) 749.00
Chemical Name or Material Azithromycin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.