Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZM 475271
GREENER_CHOICE

Catalog No. 396310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-631-0 10 mg
39-635-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-631-0 Supplier Tocris Bioscience™ Supplier No. 3963/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Src tyrosine kinase inhibitor

Src tyrosine kinase inhibitor. Reduces tumor size, vascularity and metastasis, and increases apoptosis in human pancreatic cells grown in nude mice. Also exhibits antiangiogenic activity in vitro and in vivo. Sensitizes tumor cells to the cytotoxic effects of gemcitabine (Cat. No. 3259).

Chemical Identifiers

CAS 476159-98-5
Molecular Formula C23H27ClN4O3

Specifications

Quantity 10 mg
Formula Weight 442.94
Purity >99%
Molecular Weight (g/mol) 442.944
Chemical Name or Material AZM 475271
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.