Learn More
Azotobacter vinelandii algL Polyclonal Antibody

Specifications
Specifications
| Antigen | Azotobacter vinelandii algL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 4.065 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene Alias | Alginate lyase |
| Host Species | Rabbit |
| Immunogen | Recombinant Azotobacter vinelandii Alginate lyase protein (24-374aa). |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.