Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azurocidin/CAP37/HBP Antibody (B-K37), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB340661 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340661 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340661 Supplier Novus Biologicals Supplier No. NBP314596MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Azurocidin/CAP37/HBP Monoclonal antibody specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for ELISA

Specifications

Antigen Azurocidin/CAP37/HBP
Applications ELISA
Classification Monoclonal
Clone B-K37
Conjugate mFluor Violet 450 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AZAMP, AZU, azurocidin, azurocidin 1, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin
Host Species Mouse
Immunogen Recombinant human Azurocidin/Cap37/HBP
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 566
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.