Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ B 973B

Click to view available options
Quantity:
10 mg
50 mg
Description
B 973B is a positive allosteric modulator of α7 nAChRs (EC50 ∽0.3 μM); active enantiomer. Increases potency of acetylcholine (EC50 values are 0.007 mM and 0.3 mM in the presence and absence of B 937B, respectively). Potentiates peak acetylcholine-induced currents 6-fold relative to maximal acetylcholine, and slows channel desensitization, resulting in a 6900-fold increase in charge transfer. Decreases paw edema in vivo. Analgesic.
Specifications
Specifications
Chemical Name or Material | 3,4-Difluoro-N-[(1S)-1-[6-[4-(2-pyridinyl)-1-piperazinyl]-2-pyrazinyl]ethyl]benzenepropanamide |
CAS | 2244989-34-0 |
Quantity | 10 mg |
Molecular Formula | C24H26F2N6O |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction