Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ B.O.D. Bottles, Unnumbered
SDPSCI_ED

Catalog No. p-200009369 Shop All United Scientific Supplies, Inc Products
Click to view available options
Volume (Metric):
60 mL
300 mL

BOD Bottles for incubating diluted samples of clean, waste, effluent, and polluted water during the 5-day biochemical oxygen demand (BOD) test.

  • Pack of 36 unnumbered glass BOD bottles with 60 mL capacity.
  • Ideal for incubating diluted samples of clean, waste, effluent and polluted water during the five-day biochemical oxygen demand (BOD) test.
  • Glass pennyhead stoppers included.

Specifications

Volume (Metric) 60 mL
Stopper Glass Pennyhead
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.