Learn More
Description
Specifications
Specifications
| Antigen | B-Raf |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 1H12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Gene Accession No. | P15056 |
| Gene Alias | 94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109 |
| Gene Symbols | BRAF |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
