Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-Raf Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB203511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB203511 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB203511 Supplier Novus Biologicals Supplier No. NBP241219AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

B-Raf Polyclonal antibody specifically detects B-Raf in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen B-Raf
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias 94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the center of human B-raf. The immunogen is located within amino acids 330 - 380 of B-raf. Amino Acid Squence: QPFRPADEDHRNQFGQRDR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 673
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.