Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B220/CD45R Antibody (20.96) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10065367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065367 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065367 Supplier Novus Biologicals Supplier No. NB10065367

Mouse Monoclonal Antibody

B220/CD45R Monoclonal specifically detects B220/CD45R in Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen B220/CD45R
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 20.96
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Mouse
Immunogen Ovine thymocytes
Purification Method Protein G purified
Quantity 0.125 mg
Regulatory Status RUO
Research Discipline Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Test Specificity NB100-65367 recognizes the ovine CD45R cell surface antigen, a 220kD glycoprotein, which is expressed by all B lymphocytes and a subset of T lymphocytes. In immunohistochemical staining the antibody labels all lymphocytes within the B cell follicles as well as scattered cells in interfollicular areas. *This clone has been reported to react with paraffin-embedded material following alcohol fixation. Please refer to ref. Sainte Marie, et al for alcohol fixation protocol.
Target Species Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.