Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B220/CD45R Antibody (20.96) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | B220/CD45R |
|---|---|
| Clone | 20.96 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10065367
![]() |
Novus Biologicals
NB10065367 |
0.125 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
B220/CD45R Monoclonal specifically detects B220/CD45R in Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| B220/CD45R | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Stem Cell Markers | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| PTPRC | |
| IgG1 | |
| Protein G purified | |
| NB100-65367 recognizes the ovine CD45R cell surface antigen, a 220kD glycoprotein, which is expressed by all B lymphocytes and a subset of T lymphocytes. In immunohistochemical staining the antibody labels all lymphocytes within the B cell follicles as well as scattered cells in interfollicular areas. *This clone has been reported to react with paraffin-embedded material following alcohol fixation. Please refer to ref. Sainte Marie, et al for alcohol fixation protocol. |
| 20.96 | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Sheep | |
| 5788 | |
| Ovine thymocytes | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title