Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B220/CD45R Antibody (IVA103), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB50492A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50492A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50492A594 Supplier Novus Biologicals Supplier No. NB500492AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B220/CD45R Monoclonal specifically detects B220/CD45R in Bovine samples. It is validated for Flow Cytometry.

Specifications

Antigen B220/CD45R
Applications Flow Cytometry
Classification Monoclonal
Clone IVA103
Conjugate Alexa Fluor 594
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Mouse
Immunogen Bovine leukocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Immunophenotyping, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788.0
Test Specificity The antibody IVA103 reacts with CD45R antigen (bovine), a single chain type I transmembrane glycoprotein (member of the protein tyrosine phosphatase family), a restricted form of the CD45 family. CD45R is an essential regulator of T and B lymphocytes antigen receptor signaling and it is specifically expressed in hematopoietic cells.
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.