Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B3GALNT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | B3GALNT1 |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3702
![]()
|
Proteintech Group Inc
220481AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
B3GALNT1 gene ontology annotations related to this gene include protein glycosylation.Specifications
| B3GALNT1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O75752, Q6AY39, Q920V1 | |
| 26879, 310508, 8706 | |
| B3GALNT1 Fusion Protein Ag16976 | |
| Primary | |
| -20°C | |
| B3GALNT1 |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| b3Gal T3, B3GALNT1, B3GALT3, Beta 1,3 GalNAc T1, Beta 1,3 GalTase 3, Beta 3 Gx T3, beta3Gal T3, Beta3GalT3, galT3, Gb4Cer, GLCT3, GLOB, Globoside synthase, P, P1 | |
| B3GALNT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title