Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B3GALT6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | B3GALT6 |
|---|---|
| Concentration | 0.31 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5951
![]()
|
Proteintech Group Inc
550491AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to B3GALT6.
B3GALT6, Beta-1, 3-galactosyltransferase that transfers galactose from UDP-galactose to substrates with a terminal beta-linked galactose residue. B3GALT6 has a preference for galactose-beta-1, 4-xylose that is found in the linker region of glycosaminoglycans, such as heparan sulfate and chondroitin sulfate. B3GALT6 has no activity towards substrates with terminal glucosamine or galactosamine residues.Specifications
| B3GALT6 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q91Z92, Q96L58 | |
| 117592, 126792 | |
| Peptide | |
| Primary | |
| -20°C | |
| B3GALT6 |
| 0.31 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B3GALT6, Beta 1,3 GalTase 6, Beta3Gal T6, beta3GalT6, GAG GalTII, Galactosyltransferase II | |
| B3galt6 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title