Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B3GAT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | B3GAT2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
B3GAT2 Polyclonal specifically detects B3GAT2 in Mouse samples. It is validated for Western Blot.Specifications
| B3GAT2 | |
| Polyclonal | |
| Rabbit | |
| P59270 | |
| 135152 | |
| Synthetic peptides corresponding to B3gat2 (beta-1,3-glucuronyltransferase 2 (glucuronosyltransferase S)) The peptide sequence was selected from the C terminal of B3gat2. Peptide sequence SDFLKQITTVEELEPKASNCTKVLVWHTRTEKVNLANEPKYHLDTVNIEV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Beta-1,3-glucuronyltransferase 2, beta-1,3-glucuronyltransferase 2 (glucuronosyltransferase S), EC 2.4.1.135, GlcAT-D, GlcAT-S, GLCATSgalactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 2, Glucuronosyltransferase S, KIAA1963glcAT-D, MGC138535, UDP-glucuronosyltransferase S, UDP-glucuronyltransferase S, uridine diphosphate glucuronic acid:acceptor glucuronosyltransferase | |
| B3GAT2 | |
| IgG | |
| 37 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title