Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B4GALNT3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB136135 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB136135 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB136135 Supplier Novus Biologicals Supplier No. NBP306340AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

B4GALNT3 Polyclonal specifically detects B4GALNT3 in Human samples. It is validated for Western Blot.

Specifications

Antigen B4GALNT3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias beta-1,4-N-acetyl-galactosaminyl transferase 3, beta-1,4-N-acetylgalactosaminyltransferase III, beta4GalNAcT3, N-acetyl-beta-glucosaminyl-glycoprotein 4-beta-N- acetylgalactosaminyltransferase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human B4GALNT3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283358
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.