Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B4GALT3 Antibody (OTI1G9), PE, Novus Biologicals™
SDP

Catalog No. NB288903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288903 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288903 Supplier Novus Biologicals Supplier No. NBP271386PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B4GALT3 Monoclonal antibody specifically detects B4GALT3 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen B4GALT3
Applications Western Blot
Classification Monoclonal
Clone OTI1G9
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias b4Gal-T3, beta-1,4-galactosyltransferase 3, Beta-1,4-GalTase 3, Beta4Gal-T3, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 3, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 100-393 of human B4GALT3(NP_003770) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8703
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.