Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B4GALT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7088480 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6822 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6822 Supplier Proteintech Group Inc Supplier No. 110411AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The beta-1,3 galactosyltransferase( beta3GalT) gene family encodes a set of type II transmembrane glycoproteins that are catalytically diverse and use different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine ) to catalyze the addition of an activated monosaccharide to a terminal lactose. The protein coding sequences for beta-1,3-Gal-T genes comprise a single exon and are distantly related to the Drosophila Brainiac gene. The beta-1,4-galactosyltransferase ( beta4GalT) gene family encodes type II membrane-bound glycoproteins that show exclusive specificity for the donor substrate, UDP-galactose. beta-1,4Gal-T genes transfer galactose in a beta-1,4 linkage to similar acceptor.

Specifications

Antigen B4GALT3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene B4GALT3
Gene Accession No. O60512, Q6P768, Q91YY2
Gene Alias b4Gal T3, B4GALT3, Beta 1,4 GalTase 3, beta4Gal T3
Gene Symbols B4galt3
Host Species Rabbit
Immunogen B4GALT3 Fusion Protein Ag1487
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494342, 57370, 8703
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.