Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B4GALT7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 105351AP150UL

Description
Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.Specifications
B4GALT7 | |
Polyclonal | |
Unconjugated | |
B4GALT7 | |
B4GAL T7, B4GALT7, Beta 1,4 GalTase 7, beta4Gal T7, XGALT 1, XGALT1, XGPT, XGPT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11285 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UBV7 | |
B4GALT7 | |
B4GALT7 Fusion Protein Ag0793 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction