Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B4GALT7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | B4GALT7 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6384
![]() ![]() |
Proteintech Group Inc
105351AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.Specifications
B4GALT7 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UBV7 | |
11285 | |
B4GALT7 Fusion Protein Ag0793 | |
Primary | |
-20°C | |
B4GALT7 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B4GAL T7, B4GALT7, Beta 1,4 GalTase 7, beta4Gal T7, XGALT 1, XGALT1, XGPT, XGPT1 | |
B4GALT7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title