Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-1/CD80 Antibody (16C10), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB153402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153402 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153402 Supplier Novus Biologicals Supplier No. NBP312027DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen B7-1/CD80
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 16C10
Conjugate DyLight 594
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Host Species Rabbit
Immunogen This antibody was generated by immunization of cynomolgus monkeys with human B7.1 antigen.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 941
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.