Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-1/CD80 Antibody (2740B), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB295750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295750 Supplier Novus Biologicals Supplier No. FAB7401M
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-1/CD80
Applications Flow Cytometry
Classification Monoclonal
Clone 2740B
Conjugate DyLight 594
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse B7-1/CD80, Val38-Asn246, Accession # Q00609
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 941
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.