Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-1/CD80 Antibody (RM80), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB152149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB152149 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB152149 Supplier Novus Biologicals Supplier No. NBP311977AF488
Add to Cart
Add to Cart

Rat Monoclonal Antibody

B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen B7-1/CD80
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone RM80
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Host Species Rat
Immunogen This antibody was raised by immunising Sprague Dawley rats with a mouse B cell line, BCL1 and fusing immune splenocytes with P3U1 myeloma cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 941
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.