Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Rat anti-Mouse, Clone: RM80, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP3119770.2MG
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Functional AssaySpecifications
| B7-1/CD80 | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Block/Neutralize, Functional Assay | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| This antibody was raised by immunising Sprague Dawley rats with a mouse B cell line, BCL1 and fusing immune splenocytes with P3U1 myeloma cells. | |
| 0.2 mg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
| 941 | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Functional Assay | |
| RM80 | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction