Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Rat anti-Mouse, Clone: RM80, Novus Biologicals™

Rat Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | B7-1/CD80 |
---|---|
Clone | RM80 |
Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Block/Neutralize, Functional Assay |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Functional AssaySpecifications
B7-1/CD80 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Block/Neutralize, Functional Assay | |
Unconjugated | |
RUO | |
Mouse | |
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
This antibody was raised by immunising Sprague Dawley rats with a mouse B cell line, BCL1 and fusing immune splenocytes with P3U1 myeloma cells. | |
Protein A purified |
RM80 | |
Monoclonal | |
Rat | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
PBS | |
941 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title