Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-2/CD86 Antibody (BLR029F), Novus Biologicals™
SDP

Catalog No. NBP276416 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276416 Supplier Novus Biologicals Supplier No. NBP276416
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunodiffusion.

Specifications

Antigen B7-2/CD86
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunodiffusion
Classification Monoclonal
Clone BLR029F
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 6 ul/0.5 mg lysate, Immunodiffusion 1:100 - 1:500
Formulation Borate Buffered Saline (BBS) pH 8.2 with 0.09% Sodium Azide
Gene Alias Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Gene Symbols CD86
Host Species Rabbit
Immunogen Recombinant protein representing the extracellular domain
Purification Method Antigen Affinity-purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 942.0
Target Species Human
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.