Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-2/CD86 Antibody (GL1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB174396 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174396 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174396 Supplier Novus Biologicals Supplier No. FAB741I
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Mouse samples. It is validated for CyTOF, Flow Cytometry, Neutralization, Western Blot.

Specifications

Antigen B7-2/CD86
Applications CyTOF, Flow Cytometry, Neutralization, Western Blot
Classification Monoclonal
Clone GL1
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Host Species Rat
Immunogen LPS-activated mouse B cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Dendritic Cell Markers, Diabetes Research, Immunology, Inflammation, Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 942
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.