Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H2/ICOSLG Antibody (136726), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314301 Supplier Novus Biologicals Supplier No. FAB165PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-H2/ICOSLG
Applications Flow Cytometry
Classification Monoclonal
Clone 136726
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H2, Asp19-Ser258, Accession # O75144
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23308
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.