Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H2/ICOSLG Antibody (599829), DyLight 680, Novus Biologicals™
SDP

Catalog No. MAB1581FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1581FR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1581FR Supplier Novus Biologicals Supplier No. MAB1581FR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-H2/ICOSLG
Applications Flow Cytometry
Classification Monoclonal
Clone 599829
Conjugate DyLight 680
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Gene Symbols ICOSLG
Host Species Rat
Immunogen rmB7-H2 aa 47-279
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23308
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.