Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H2/ICOSLG Antibody (599841), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB349782 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349782 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349782 Supplier Novus Biologicals Supplier No. FAB158MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

B7-H2/ICOSLG Monoclonal antibody specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen B7-H2/ICOSLG
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 599841
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse B7-H2 , Glu47-Lys279, Accession # Q9JHJ8
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23308
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.