Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H3/CD276 Antibody (185504), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB315279 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315279 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315279 Supplier Novus Biologicals Supplier No. FAB1027PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-H3/CD276
Applications Flow Cytometry
Classification Monoclonal
Clone 185504
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H3, Leu29-Pro245, Accession # NP_079516
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 80381
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.