Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H3/CD276 Antibody (317), Alexa Fluor™ 350
SDP

Catalog No. NB077423 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB077423 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB077423 Supplier Novus Biologicals Supplier No. NBP290756AF350

Rabbit Monoclonal Antibody

B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Mouse samples. It is validated for ELISA.

Specifications

Antigen B7-H3/CD276
Applications ELISA
Classification Monoclonal
Clone 317
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse B7-H3/CD276 (NP_598744.1; Met1-Phe244).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 80381
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.