Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H3/CD276 Antibody (MJ18) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP275194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275194 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275194 Supplier Novus Biologicals Supplier No. NBP275194
Add to Cart
Add to Cart

Rat Monoclonal Antibody

B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Block/Neutralize.

Specifications

Antigen B7-H3/CD276
Applications Flow Cytometry, ELISA, Immunofluorescence, Neutralization
Classification Monoclonal
Clone MJ18
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, ELISA, Immunofluorescence, Neutralization
Formulation PBS with 0.02% Proclin 300
Gene Alias B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule
Gene Symbols CD276
Host Species Rat
Immunogen This antibody was generated by immunizing Sprague Dawley rats with B7-H3-Ig, consisting of the extracellular domain (aa1-242 of mouse B7-H3) linked to the Fc portion of mouse IgG2a, emulsified in CFA (Difco). The antibody was selected by its reactivity to mouse B7-H3-transfected cells, but not to parental cells by flow cytometry.
Purification Method Protein A purified
Quantity 0.2 mg
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 80381.0
Test Specificity This antibody binds to mouse B7-H3 and does not react with other B7-family molecules.
Target Species Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.