Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H3/CD276 Antibody (MM0104-20J12) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019018 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019018 0.025 mg
NBP212048 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019018 Supplier Novus Biologicals Supplier No. NBP2120480.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot.

Specifications

Antigen B7-H3/CD276
Applications Western Blot
Classification Monoclonal
Clone MM0104-20J12
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. NP_079516
Gene Alias B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule
Gene Symbols CD276
Host Species Mouse
Immunogen Human CD276
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80381
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.