Learn More
Description
Specifications
Specifications
| Antigen | B7-H4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 188 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry |
| Gene Alias | B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1 |
| Gene Symbols | VTCN1 |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was Purified B7H4. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
