Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H4 Antibody (188), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP143266IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143266IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143266IR Supplier Novus Biologicals Supplier No. NBP143266IR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

B7-H4 Monoclonal specifically detects B7-H4 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-H4
Applications Flow Cytometry
Classification Monoclonal
Clone 188
Conjugate DyLight 755
Dilution Flow Cytometry
Gene Alias B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1
Gene Symbols VTCN1
Host Species Rat
Immunogen The immunogen for this antibody was Purified B7H4.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 79679
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.