Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H4 Antibody (2318A), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311599 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311599 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311599 Supplier Novus Biologicals Supplier No. FAB65764PEATT594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

B7-H4 Monoclonal antibody specifically detects B7-H4 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen B7-H4
Applications Flow Cytometry
Classification Monoclonal
Clone 2318A
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human B7-H4, Phe29-Ser258, Accession # Q7Z7D3
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 79679
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.