Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H6 Antibody (875001), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB375069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375069 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375069 Supplier Novus Biologicals Supplier No. FAB7144M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Gene Knock-Out.

Specifications

Antigen B7-H6
Applications CyTOF, ELISA, Flow Cytometry, Gene Knock-Out
Classification Monoclonal
Clone 875001
Conjugate DyLight 594
Dilution Flow Cytometry, ELISA, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias B7 homolog 6, B7-H6, DKFZp686O24166
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H6, Met1-Ser262, Accession # Q68D85
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 374383
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.