Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H6 Antibody (875023), DyLight 350, R&D Systems™
SDP

Catalog No. MAB71441UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB71441UV 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB71441UV Supplier Novus Biologicals Supplier No. MAB71441UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining).

Specifications

Antigen B7-H6
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 875023
Conjugate DyLight 350
Dilution Flow Cytometry, Flow (Intracellular)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B7 homolog 6, B7-H6, DKFZp686O24166
Gene Symbols NCR3LG1
Host Species Mouse
Immunogen Partial recombinant human B7-H6 protein (amino acids1-262). [UniProt Q68D85]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374383
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.