Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H6 Antibody (B7H6/4821), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB381372 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381372 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381372 Supplier Novus Biologicals Supplier No. NBP308339R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen B7-H6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone B7H6/4821
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias B7 homolog 6, B7-H6, DKFZp686O24166
Host Species Mouse
Immunogen Recombinant fragment (around aa38-169) of human B7-H6 protein (exact sequence is proprietary) (Uniprot: Q68D85 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 374383
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.