Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H7/HHLA2 Antibody (907812R), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346324 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346324 Supplier Novus Biologicals Supplier No. FAB80841RMFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H7/HHLA2 Monoclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry.

Specifications

Antigen B7-H7/HHLA2
Applications Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry
Classification Monoclonal
Clone 907812R
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, Human endogenous retrovirus-H long terminal repeat-associating protein 2
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2, Met1-Asn344, Accession # Q9UM44
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Inflammation
Primary or Secondary Primary
Gene ID (Entrez) 11148
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.