Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H7/HHLA2 Antibody (907834) - Azide and BSA Free, R&D Systems™
SDP

Catalog No. MAB80842SP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB80842SP 25 μg
MAB80842 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB80842SP Supplier Novus Biologicals Supplier No. MAB80842SP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H7/HHLA2 Monoclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen B7-H7/HHLA2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 907834
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, Human endogenous retrovirus-H long terminal repeat-associating protein 2
Gene Symbols HHLA2
Host Species Mouse
Immunogen Partial recombinant human B7-H7 protein (amino acids 1-344) [UniProt Q9UM44]
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Immunology, Inflammation
Primary or Secondary Primary
Gene ID (Entrez) 11148
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.