Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H7/HHLA2 Antibody (907834), Biotin, R&D Systems™
SDP

Catalog No. MAB80842B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB80842B 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB80842B Supplier Novus Biologicals Supplier No. MAB80842B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B7-H7/HHLA2 Monoclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-H7/HHLA2
Applications Flow Cytometry
Classification Monoclonal
Clone 907834
Conjugate Biotin
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, Human endogenous retrovirus-H long terminal repeat-associating protein 2
Gene Symbols HHLA2
Host Species Mouse
Immunogen Partial recombinant human B7-H7 protein (amino acids 1-344) [UniProt Q9UM44]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11148
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.