Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human B9D1 His Protein
SDP

Catalog No. NBP25181505 Shop All Novus Biologicals Products
Click to view available options
Quantity:
0.1 mg
0.5 mg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-204 of Human B9D1 The Recombinant Human B9D1 Protein is derived from E. coli. The Recombinant Human B9D1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 0.4M UREA
Gene ID (Entrez) 27077
Molecular Weight (g/mol) TMW: 25.2kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias B9, B9 domain-containing protein 1, B9 protein domain 1, endothelial precursor protein B9, EPPB9, MKS1-related protein 1, MKSR1
Common Name B9D1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.