Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BA Antigen (Leu21) Antibody (DF1588), Novus Biologicals™
SDP

Catalog No. NB253349200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253349200 200 μg
NB25334920 20 μg
NB253349100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253349200 Supplier Novus Biologicals Supplier No. NBP253349200UG

Mouse Monoclonal Antibody

BA Antigen (Leu21) Monoclonal antibody specifically detects BA Antigen (Leu21) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, SDS-Page.

Specifications

Antigen BA Antigen (Leu21)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page
Classification Monoclonal
Clone DF1588
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Host Species Mouse
Immunogen Human leucocytes
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.