Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAAT1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215536 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215536 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215536 Supplier Novus Biologicals Supplier No. NBP215536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAAT1 Polyclonal specifically detects BAAT1 in Human samples. It is validated for Western Blot.

Specifications

Antigen BAAT1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q6PJG6
Gene Alias BAAT1, BRCA1-associated ATM activator 1, BRCA1-associated protein required for ATM activation protein 1, C7orf27, chromosome 7 open reading frame 27, HEAT repeat-containing protein C7orf27, MGC22916
Gene Symbols BRAT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human BRAT1. The exact sequence is proprietary.
Molecular Weight of Antigen 88 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 221927
Test Specificity Human. Expected to cross react based on sequence identity: Bovine (80%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.