Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baboon Antibody, Novus Biologicals™
SDP

Catalog No. NB018168 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018168 0.025 mg
NB300665005 0.05 mg
NB300665 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018168 Supplier Novus Biologicals Supplier No. NB3006650.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Baboon Polyclonal specifically detects Baboon in Drosophila samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Baboon
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200, Immunoprecipitation 1:200
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias Activin A receptor 45A, ATR 1, Atr 45A, Atr I, ATR1, Atr45A, AtrI, Bab, Babo, CG8224, G protein coupled receptor kinase 3, Gprk 3, Gprk3, l(2)k16912, RE55648p, STK E, STKE
Host Species Rabbit
Immunogen Synthetic peptide: QPDPSIEEMKKVVCIEKCRPN, corresponding to amino acids 552 to 572 of Baboon.The was generated against a peptide sequence near the C terminal end of the protein.The peptides was amidated for conjugation.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Test Specificity The is unique to Baboon protein.
Target Species Drosophila
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.